Product Details

SNP ID
rs2720575
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.8:17578573 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
CTCCTGGCAGGCCCTTGTGTGCAAC[C/T]GCTGAAAAGGCAGAAATGTAATTTG
Phenotype
MIM: 604584 MIM: 601872
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PDGFRL PubMed Links

Gene Details

Gene
PDGFRL
Gene Name
platelet derived growth factor receptor like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006207.2 Intron NP_006198.1
Gene
SLC7A2
Gene Name
solute carrier family 7 member 2
There are no transcripts associated with this gene.

View Full Product Details