Product Details

SNP ID
rs2332655
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:173171851 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAACCACACAGCAATCAAACCTC[A/G]ATAAATATATAAATAAAAATTGTGC
Phenotype
MIM: 605005
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GALNT7 PubMed Links

Gene Details

Gene
GALNT7
Gene Name
polypeptide N-acetylgalactosaminyltransferase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017423.2 Intron NP_059119.2
XM_005263062.1 Intron XP_005263119.1
XM_011532025.2 Intron XP_011530327.1
XM_017008292.1 Intron XP_016863781.1
XM_017008293.1 Intron XP_016863782.1
Gene
LOC101930370
Gene Name
uncharacterized LOC101930370
There are no transcripts associated with this gene.

View Full Product Details