Product Details

SNP ID
rs6427323
Assay Type
Functionally tested
NCBI dbSNP Submissions
37
Location
Chr.1:156607997 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGATTCAATCCTTAGCCTAGGCAG[C/G]TTCTAATTTTCTGACTTCATTAACC
Phenotype
MIM: 608862
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GPATCH4 PubMed Links

Gene Details

Gene
GPATCH4
Gene Name
G-patch domain containing 4
There are no transcripts associated with this gene.

Gene
HAPLN2
Gene Name
hyaluronan and proteoglycan link protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021817.2 Intron NP_068589.1
XM_005245415.2 Intron XP_005245472.1
XM_011509853.2 Intron XP_011508155.1
XM_017002020.1 Intron XP_016857509.1
XM_017002021.1 Intron XP_016857510.1
Gene
LOC101928177
Gene Name
uncharacterized LOC101928177
There are no transcripts associated with this gene.

Gene
NAXE
Gene Name
NAD(P)HX epimerase
There are no transcripts associated with this gene.

View Full Product Details