Product Details
- SNP ID
-
rs6062690
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:62648884 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCGAGTGACCGCCCCTGGGCTCCTC[G/T]TGGGTGCGACAGTGTCCTGGGAGTC
- Phenotype
-
MIM: 612436
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SLCO4A1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79685580] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SLCO4A1
- Gene Name
- solute carrier organic anion transporter family member 4A1
View Full Product Details