Product Details

SNP ID
rs6685250
Assay Type
Functionally tested
NCBI dbSNP Submissions
28
Location
Chr.1:31261901 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCTTCTGAAATGGGAAACACCACA[G/T]GGAAGAAAAGAAAGTTTTTCCTGCA
Phenotype
MIM: 607797
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SNRNP40 PubMed Links

Gene Details

Gene
SNRNP40
Gene Name
small nuclear ribonucleoprotein U5 subunit 40
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004814.2 Intron NP_004805.2

View Full Product Details