Product Details
- SNP ID
-
rs3094478
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:2697049 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CACCGGTCTGCCTCTATCCCGCCCC[A/G]ATGCAGCCACTCACGCCCTGGGTGC
- Phenotype
-
MIM: 611285
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
KCTD5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78206812] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KCTD5
- Gene Name
- potassium channel tetramerization domain containing 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018992.3 |
|
Intron |
|
|
NP_061865.1 |
View Full Product Details