Product Details
- SNP ID
-
rs7999481
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:53031025 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AACCAGCATTTGGGTAAGATTATCA[A/G]CTATAATAATAGAAAATAAAATACA
- Phenotype
-
MIM: 614061
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
OLFM4
PubMed Links
Gene Details
- Gene
- OLFM4
- Gene Name
- olfactomedin 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006418.4 |
|
Intron |
|
|
NP_006409.3 |
View Full Product Details