Product Details

SNP ID
rs10711393
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:56653147 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAGTGTTAAATATTGGTTCCAAA[A/T]AATGAAATTATGTTTCTTCTCAAAT
Phenotype
MIM: 600497
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC101929935 PubMed Links

Gene Details

Gene
LOC101929935
Gene Name
uncharacterized LOC101929935
There are no transcripts associated with this gene.

Gene
PRKAA2
Gene Name
protein kinase AMP-activated catalytic subunit alpha 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006252.3 Intron NP_006243.2
XM_017001692.1 Intron XP_016857181.1
XM_017001693.1 Intron XP_016857182.1
XM_017001694.1 Intron XP_016857183.1

View Full Product Details