Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001321080.1 | Intron | NP_001308009.1 | |||
NM_001321083.1 | Intron | NP_001308012.1 | |||
NM_001321084.1 | Intron | NP_001308013.1 | |||
NM_020771.3 | Intron | NP_065822.2 | |||
XM_017011113.1 | Intron | XP_016866602.1 | |||
XM_017011114.1 | Intron | XP_016866603.1 | |||
XM_017011115.1 | Intron | XP_016866604.1 | |||
XM_017011116.1 | Intron | XP_016866605.1 | |||
XM_017011118.1 | Intron | XP_016866607.1 | |||
XM_017011119.1 | Intron | XP_016866608.1 | |||
XM_017011120.1 | Intron | XP_016866609.1 | |||
XM_017011121.1 | Intron | XP_016866610.1 | |||
XM_017011122.1 | Intron | XP_016866611.1 |