Product Details

SNP ID
rs1344279
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:121896832 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGGCCCAGGAGTTCAAGGTTATCA[A/G]GCACCTGTAATTGTGCCAGTGAATG
Phenotype
MIM: 602339
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
SLC15A2 PubMed Links

Gene Details

Gene
SLC15A2
Gene Name
solute carrier family 15 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145998.1 Intron NP_001139470.1
NM_021082.3 Intron NP_066568.3
XM_005247722.3 Intron XP_005247779.1
XM_006713736.3 Intron XP_006713799.1
XM_017007074.1 Intron XP_016862563.1

View Full Product Details