Product Details
- SNP ID
-
rs12777477
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:5870180 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TCCACATCCCTCTCCACTGCTTCAT[C/T]TGCAAGCTCCAGGTTCTGCCTTCCC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ANKRD16
PubMed Links
Gene Details
- Gene
- ANKRD16
- Gene Name
- ankyrin repeat domain 16
View Full Product Details