Product Details

SNP ID
rs1885673
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:15257104 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGCTGGGGCATGAAATCCAGGTCC[C/T]GATACAGTAGAAGGCCCTCTCTGTT
Phenotype
MIM: 601594
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
JARID2 PubMed Links

Gene Details

Gene
JARID2
Gene Name
jumonji and AT-rich interaction domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267040.1 Intron NP_001253969.1
NM_004973.3 Intron NP_004964.2
XM_005249089.3 Intron XP_005249146.1
XM_011514579.2 Intron XP_011512881.1
XM_011514584.2 Intron XP_011512886.1
XM_017010833.1 Intron XP_016866322.1
XM_017010834.1 Intron XP_016866323.1
XM_017010835.1 Intron XP_016866324.1
Gene
JARID2-AS1
Gene Name
JARID2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details