Product Details

SNP ID
rs538779
Assay Type
Functionally tested
NCBI dbSNP Submissions
65
Location
Chr.1:111759597 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGATAACAATGCCTGTTGGTAACCA[A/G]GGCTTTTAAGGAACTACACACTTTT
Phenotype
MIM: 606168
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DDX20 PubMed Links

Gene Details

Gene
DDX20
Gene Name
DEAD-box helicase 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007204.4 Intron NP_009135.4
Gene
FAM212B
Gene Name
family with sequence similarity 212 member B
There are no transcripts associated with this gene.

Gene
LOC101928718
Gene Name
uncharacterized LOC101928718
There are no transcripts associated with this gene.

View Full Product Details