Product Details
- SNP ID
-
rs8480
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
80
- Location
-
Chr.1:151760859 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTCGCCCCACCGTGTGATAGGCTCT[G/T]CTGGTAACTTTAATGTATGTGGGGC
- Phenotype
-
MIM: 611824
MIM: 605138
MIM: 609501
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MRPL9
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74228558] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MRPL9
- Gene Name
- mitochondrial ribosomal protein L9
- Gene
- OAZ3
- Gene Name
- ornithine decarboxylase antizyme 3
There are no transcripts associated with this gene.
- Gene
- TDRKH
- Gene Name
- tudor and KH domain containing
There are no transcripts associated with this gene.
View Full Product Details