Product Details

SNP ID
rs7926767
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:123906829 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCATCTGCAGCCCACTGCTCTACA[A/G]GGTCATCATGTCCCCTAGGGTCTGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR8D4 PubMed Links

Gene Details

Gene
OR8D4
Gene Name
olfactory receptor family 8 subfamily D member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005197.1 398 Missense Mutation AAG,AGG K133R NP_001005197.1

View Full Product Details