Product Details

SNP ID
rs11946367
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:99552960 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAAACAAAATTGTTTTAGTTCAGA[A/G]AAACATAAACATGAGAAAAAATGTG
Phenotype
MIM: 616013
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRMT10A PubMed Links

Gene Details

Gene
TRMT10A
Gene Name
tRNA methyltransferase 10A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134665.2 Intron NP_001128137.1
NM_001134666.2 Intron NP_001128138.1
NM_152292.4 Intron NP_689505.1
XM_005263352.3 Intron XP_005263409.1
XM_006714417.2 Intron XP_006714480.1

View Full Product Details