Product Details

SNP ID
rs2035650
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.16:2345050 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTCCATGGTCATAGTTTGGTAAATA[A/C]GCTTCTTAAAGTTTTTTAAATGTAT
Phenotype
MIM: 601615
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ABCA17P PubMed Links

Gene Details

Gene
ABCA17P
Gene Name
ATP binding cassette subfamily A member 17, pseudogene
There are no transcripts associated with this gene.

Gene
ABCA3
Gene Name
ATP binding cassette subfamily A member 3
There are no transcripts associated with this gene.

View Full Product Details