Product Details
- SNP ID
-
rs2025717
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:15080778 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GATCTTTATCCTTTTCCCTTTCTAA[A/C]CTCTTAGTTTATACTCTACAAAGAG
- Phenotype
-
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ACBD7
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11259467] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ACBD7
- Gene Name
- acyl-CoA binding domain containing 7
- Gene
- OLAH
- Gene Name
- oleoyl-ACP hydrolase
There are no transcripts associated with this gene.
View Full Product Details