Product Details
- SNP ID
-
rs1916289
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:49255909 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGAATGTTTGTGATGTGGGCGATTC[C/T]GACTGAGGGGGTCTGTAGACCACAT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FLJ40194
PubMed Links
Gene Details
- Gene
- FLJ40194
- Gene Name
- uncharacterized FLJ40194
There are no transcripts associated with this gene.
View Full Product Details