Product Details

SNP ID
rs1950189
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:28781296 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTGCCTTCTGACTGGAGCCCTGCT[A/G]GTGAGTGGGCAAGTTGTCCTATTTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC01551 PubMed Links

Gene Details

Gene
LINC01551
Gene Name
long intergenic non-protein coding RNA 1551
There are no transcripts associated with this gene.

Gene
LOC105370424
Gene Name
uncharacterized LOC105370424
There are no transcripts associated with this gene.

View Full Product Details