Product Details

SNP ID
rs1883424
Assay Type
Functionally tested
NCBI dbSNP Submissions
26
Location
Chr.1:1716489 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTAGCATGCTTCCCTAATGAGAA[A/G]TAAAGTGTCATGCAAAGAAACCTCA
Phenotype
MIM: 116951
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDK11A PubMed Links

Gene Details

Gene
CDK11A
Gene Name
cyclin dependent kinase 11A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001313896.1 Intron NP_001300825.1
NM_001313982.1 Intron NP_001300911.1
NM_024011.3 Intron NP_076916.2
NM_033529.3 Intron NP_277071.2
Gene
SLC35E2
Gene Name
solute carrier family 35 member E2
There are no transcripts associated with this gene.

View Full Product Details