Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001317188.1 | Intron | NP_001304117.1 | |||
NM_199051.2 | Intron | NP_950252.1 | |||
XM_011509472.2 | Intron | XP_011507774.1 | |||
XM_011509475.2 | Intron | XP_011507777.1 | |||
XM_011509476.2 | Intron | XP_011507778.1 | |||
XM_017001125.1 | Intron | XP_016856614.1 | |||
XM_017001126.1 | Intron | XP_016856615.1 | |||
XM_017001127.1 | Intron | XP_016856616.1 | |||
XM_017001128.1 | Intron | XP_016856617.1 | |||
XM_017001129.1 | Intron | XP_016856618.1 | |||
XM_017001130.1 | Intron | XP_016856619.1 | |||
XM_017001131.1 | Intron | XP_016856620.1 | |||
XM_017001132.1 | Intron | XP_016856621.1 | |||
XM_017001133.1 | Intron | XP_016856622.1 |