Product Details
- SNP ID
-
rs1966426
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:201703196 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCCAGAAACCTGACATTGAATTCTA[A/G]TTATTTTTCTTATTTTAATTTACCT
- Phenotype
-
MIM: 606352
MIM: 606575
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ALS2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs202075262] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ALS2
- Gene Name
- ALS2, alsin Rho guanine nucleotide exchange factor
- Gene
- MPP4
- Gene Name
- membrane palmitoylated protein 4
There are no transcripts associated with this gene.
View Full Product Details