Product Details
- SNP ID
-
rs6572710
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:50907707 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGTCTGCTACAGAGTGGGTGCTCAA[A/T]AAATATTGAATTATTTGTGACTTTT
- Phenotype
-
MIM: 613741
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ABHD12B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs199705432] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ABHD12B
- Gene Name
- abhydrolase domain containing 12B
There are no transcripts associated with this gene.
- Gene
- PYGL
- Gene Name
- phosphorylase, glycogen, liver
View Full Product Details