Product Details

SNP ID
rs5154
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44943828 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATTCTCACGATAACCCTGAGGTA[C/G]ATATTATTACCCCGTTCTACAAAAG
Phenotype
MIM: 608083 MIM: 600745
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
APOC2 PubMed Links

Gene Details

Gene
APOC2
Gene Name
apolipoprotein C2
There are no transcripts associated with this gene.

Gene
APOC4
Gene Name
apolipoprotein C4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001646.2 Intron NP_001637.1
Gene
APOC4-APOC2
Gene Name
APOC4-APOC2 readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details