Product Details

SNP ID
rs1064725
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44919304 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCAGCGCCTGTGCTGAGGACTCCC[G/T]CCATGTGGCCCCAGGTGCCACCAAT
Phenotype
MIM: 107710 MIM: 107741
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
APOC1 PubMed Links

Gene Details

Gene
APOC1
Gene Name
apolipoprotein C1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321065.1 525 UTR 3 NP_001307994.1
NM_001321066.1 525 UTR 3 NP_001307995.1
NM_001645.4 525 UTR 3 NP_001636.1
Gene
APOC1P1
Gene Name
apolipoprotein C1 pseudogene 1
There are no transcripts associated with this gene.

Gene
APOE
Gene Name
apolipoprotein E
There are no transcripts associated with this gene.

View Full Product Details