Product Details

SNP ID
rs1869447
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:69052600 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCAGTGCAGCAGGGCCGGGCAGAG[C/T]GAGGGTGGGGTCCGGTCTCCTGCCT
Phenotype
MIM: 612163
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TPCN2 PubMed Links

Gene Details

Gene
TPCN2
Gene Name
two pore segment channel 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139075.3 Intron NP_620714.2
XM_005273824.3 Intron XP_005273881.1
XM_005273826.3 Intron XP_005273883.1
XM_005273830.3 Intron XP_005273887.1
XM_005273831.3 Intron XP_005273888.1
XM_005273832.3 Intron XP_005273889.1
XM_011544802.2 Intron XP_011543104.1
XM_011544805.1 Intron XP_011543107.1
XM_011544806.1 Intron XP_011543108.1
XM_011544807.2 Intron XP_011543109.1
XM_011544808.2 Intron XP_011543110.1
XM_017017328.1 Intron XP_016872817.1
XM_017017329.1 Intron XP_016872818.1
XM_017017330.1 Intron XP_016872819.1
XM_017017331.1 Intron XP_016872820.1
XM_017017332.1 Intron XP_016872821.1
XM_017017333.1 Intron XP_016872822.1
XM_017017334.1 Intron XP_016872823.1
XM_017017335.1 Intron XP_016872824.1
XM_017017336.1 Intron XP_016872825.1

View Full Product Details