Product Details

SNP ID
rs1805362
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:94420160 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTCTTCTTAAAGAACTAGTGTTCA[C/T]AAAAGGATCATCATCATCATCCTGA
Phenotype
MIM: 600814
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MRE11A PubMed Links

Gene Details

Gene
MRE11A
Gene Name
MRE11 homolog A, double strand break repair nuclease
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005590.3 1996 Missense Mutation ATG,GTG M670V NP_005581.2
NM_005591.3 1996 Missense Mutation ATG,GTG M698V NP_005582.1
XM_005274008.3 1996 Missense Mutation ATG,GTG M542V XP_005274065.1
XM_006718842.3 1996 Missense Mutation ATG,GTG M697V XP_006718905.1
XM_011542837.2 1996 Missense Mutation ATG,GTG M698V XP_011541139.1
XM_017017772.1 1996 Missense Mutation ATG,GTG M698V XP_016873261.1
XM_017017773.1 1996 Missense Mutation ATG,GTG M697V XP_016873262.1

View Full Product Details