Product Details

SNP ID
rs11082781
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.18:49788394 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAAAGGCATAATGAAACCTAAAT[A/G]TAATATTTATTTTGTCACGTAGTTA
Phenotype
MIM: 604770
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ACAA2 PubMed Links
Additional Information
For this assay, SNP(s) [rs114836888] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
ACAA2
Gene Name
acetyl-CoA acyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006111.2 Intron NP_006102.2

View Full Product Details