Product Details

SNP ID
rs9916983
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:40931825 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCACAGGGAGGGCTTGGGGTAAAT[A/T]GTTTGTTTTGTTGTTGTTGTTGTTG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CYP2B7P PubMed Links

Gene Details

Gene
CYP2B7P
Gene Name
cytochrome P450 family 2 subfamily B member 7, pseudogene
There are no transcripts associated with this gene.

View Full Product Details