Product Details
- SNP ID
-
rs5999962
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:35709608 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGTGCTCTTTTATATCTAGAAATA[C/T]TTCTTCAATTTTATCTTTAAATATT
- Phenotype
-
MIM: 607255
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
APOL5
PubMed Links
Gene Details
- Gene
- APOL5
- Gene Name
- apolipoprotein L5
View Full Product Details