Product Details

SNP ID
rs4973245
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:230039159 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTGATGAATATGCCTGGATTATCT[A/G]TTTACAGGGATCAGTCAAGACATTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC16A14 PubMed Links

Gene Details

Gene
SLC16A14
Gene Name
solute carrier family 16 member 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152527.4 Intron NP_689740.2
XM_005246353.3 Intron XP_005246410.1
XM_011510751.2 Intron XP_011509053.1
XM_011510752.1 Intron XP_011509054.1
XM_017003480.1 Intron XP_016858969.1

View Full Product Details