Product Details

SNP ID
rs11880020
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:52299547 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGATTTTATTAATTTGTTAGCCTC[A/G]TTCACAGAACTCAAGGAAGCATTTT
Phenotype
MIM: 613910
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF480 PubMed Links

Gene Details

Gene
ZNF480
Gene Name
zinc finger protein 480
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297624.1 Intron NP_001284553.1
NM_001297625.1 Intron NP_001284554.1
NM_144684.3 Intron NP_653285.2
XM_011526465.2 Intron XP_011524767.1
Gene
ZNF766
Gene Name
zinc finger protein 766
There are no transcripts associated with this gene.

View Full Product Details