Product Details
- SNP ID
-
rs13010821
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:240476621 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CAGCAGAGCACAGAAGGGGCCCCTA[C/T]GGCAACTAAGAGGCAGGCATATGCC
- Phenotype
-
MIM: 600395
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ANKMY1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs80086246] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ANKMY1
- Gene Name
- ankyrin repeat and MYND domain containing 1
- Gene
- GPC1
- Gene Name
- glypican 1
There are no transcripts associated with this gene.
View Full Product Details