Product Details

SNP ID
rs2314331
Assay Type
Functionally tested
NCBI dbSNP Submissions
75
Location
Chr.1:19648175 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
GCTAATCCACTCTACAAAGGGTTTC[A/G]CGGTGGGCGGGAGTCAGGAAGAAGT
Phenotype
MIM: 600613
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MINOS1-NBL1 PubMed Links

Gene Details

Gene
MINOS1-NBL1
Gene Name
MINOS1-NBL1 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204088.1 Intron NP_001191017.1
NM_001204089.1 Intron NP_001191018.1
Gene
NBL1
Gene Name
neuroblastoma 1, DAN family BMP antagonist
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204084.2 Intron NP_001191013.1
NM_001204085.1 Intron NP_001191014.1
NM_001204086.2 Intron NP_001191015.1
NM_001278164.1 Intron NP_001265093.1
NM_001278165.1 Intron NP_001265094.1
NM_001278166.1 Intron NP_001265095.1
NM_005380.7 Intron NP_005371.2
NM_182744.3 Intron NP_877421.2

View Full Product Details