Product Details

SNP ID
rs6906678
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.6:151451260 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTCTCTTATCTTCCTCCACTTTGCA[A/T]TTCTCCATTGTTTAACATCACTTGC
Phenotype
MIM: 616332 MIM: 614917
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ARMT1 PubMed Links

Gene Details

Gene
ARMT1
Gene Name
acidic residue methyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286562.1 Intron NP_001273491.1
NM_024573.2 Intron NP_078849.1
Gene
RMND1
Gene Name
required for meiotic nuclear division 1 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271937.1 Intron NP_001258866.1
NM_017909.3 Intron NP_060379.2
XM_005267040.3 Intron XP_005267097.1
XM_017010988.1 Intron XP_016866477.1

View Full Product Details