Product Details

SNP ID
rs2736980
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:87610715 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACATTAGTGCTGAGCCTGGTGATG[C/T]CCCCATAACCACACCAGGTATGTTC
Phenotype
MIM: 125485
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DSPP PubMed Links

Gene Details

Gene
DSPP
Gene Name
dentin sialophosphoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014208.3 Intron NP_055023.2

View Full Product Details