Product Details

SNP ID
rs11713574
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:11143652 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCCCTCCAGTCTCCCTTGTAGAG[G/T]AGTAGATGAATGGTGACCCCTGGTC
Phenotype
MIM: 600167
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HRH1 PubMed Links

Gene Details

Gene
HRH1
Gene Name
histamine receptor H1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000861.3 Intron NP_000852.1
NM_001098211.1 Intron NP_001091681.1
NM_001098212.1 Intron NP_001091682.1
NM_001098213.1 Intron NP_001091683.1
XM_011533652.1 Intron XP_011531954.1
XM_011533653.2 Intron XP_011531955.1
XM_017006283.1 Intron XP_016861772.1
XM_017006284.1 Intron XP_016861773.1

View Full Product Details