Product Details

SNP ID
rs10231604
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100148578 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACCTGGGAAGGGATTCATTTGGGG[A/G]GGTCACACCAGCGGGCATCCTGGCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C7orf43 PubMed Links

Gene Details

Gene
C7orf43
Gene Name
chromosome 7 open reading frame 43
There are no transcripts associated with this gene.

Gene
LAMTOR4
Gene Name
late endosomal/lysosomal adaptor, MAPK and MTOR activator 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008395.3 Intron NP_001008396.1
NM_001318236.1 Intron NP_001305165.1
NM_001318237.1 Intron NP_001305166.1
XM_017012198.1 Intron XP_016867687.1
XM_017012199.1 Intron XP_016867688.1
Gene
MIR4658
Gene Name
microRNA 4658
There are no transcripts associated with this gene.

View Full Product Details