Product Details

SNP ID
rs11606443
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:8022374 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCCCATCCTAACTCTATAAGGAA[A/C]CATTGTTAATAGTTAACTTTCACTT
Phenotype
MIM: 601197
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CASC23 PubMed Links

Gene Details

Gene
CASC23
Gene Name
cancer susceptibility candidate 23 (non-protein coding)
There are no transcripts associated with this gene.

Gene
TUB
Gene Name
tubby bipartite transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003320.4 Intron NP_003311.2
NM_177972.2 Intron NP_813977.1
XM_005253109.3 Intron XP_005253166.1
XM_011520344.2 Intron XP_011518646.1

View Full Product Details