Product Details

SNP ID
rs10769932
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:8614792 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATGGCAGGGACTGCCATGGAACA[C/G]AGCATATGGAAGGTATGGTTGCCAC
Phenotype
MIM: 612000
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TRIM66 PubMed Links

Gene Details

Gene
TRIM66
Gene Name
tripartite motif containing 66
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014818.1 7852 UTR 3 NP_055633.1
XM_006718397.2 7852 UTR 3 XP_006718460.1
XM_006718398.3 7852 UTR 3 XP_006718461.1
XM_011520504.1 7852 UTR 3 XP_011518806.1
XM_011520507.2 7852 UTR 3 XP_011518809.1
XM_011520508.1 7852 UTR 3 XP_011518810.1
XM_011520509.1 7852 UTR 3 XP_011518811.1
XM_011520510.1 7852 UTR 3 XP_011518812.1
XM_011520511.1 7852 UTR 3 XP_011518813.1
XM_011520512.1 7852 UTR 3 XP_011518814.1
XM_011520513.1 7852 UTR 3 XP_011518815.1
XM_011520514.2 7852 UTR 3 XP_011518816.1
XM_011520515.1 7852 UTR 3 XP_011518817.1
XM_011520516.1 7852 UTR 3 XP_011518818.1
XM_011520517.2 7852 UTR 3 XP_011518819.1
XM_011520518.1 7852 UTR 3 XP_011518820.1
XM_011520519.1 7852 UTR 3 XP_011518821.1
XM_011520522.2 7852 UTR 3 XP_011518824.1
XM_011520523.2 7852 UTR 3 XP_011518825.1
XM_011520524.1 7852 Intron XP_011518826.1
XM_011520525.1 7852 Intron XP_011518827.1
XM_011520526.2 7852 UTR 3 XP_011518828.1
XM_011520527.1 7852 UTR 3 XP_011518829.1
XM_017018629.1 7852 UTR 3 XP_016874118.1
XM_017018630.1 7852 UTR 3 XP_016874119.1
XM_017018631.1 7852 UTR 3 XP_016874120.1

View Full Product Details