Product Details

SNP ID
rs10823268
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:68907729 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGTTGAATCCTTGTGTTATGGCC[C/T]TTTTTTTTTTAATTTGTGAATTTGC
Phenotype
MIM: 610373
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DDX50 PubMed Links

Gene Details

Gene
DDX50
Gene Name
DEAD-box helicase 50
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024045.1 Intron NP_076950.1
XM_005270148.2 Intron XP_005270205.1
XM_011540144.2 Intron XP_011538446.1
XM_017016626.1 Intron XP_016872115.1
XM_017016627.1 Intron XP_016872116.1
XM_017016628.1 Intron XP_016872117.1
XM_017016629.1 Intron XP_016872118.1
XM_017016630.1 Intron XP_016872119.1

View Full Product Details