Product Details

SNP ID
rs1956367
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:59504450 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCTTAACAGTGGCGTAATTGTAC[G/T]TACCTGTTGTGTTTCAGTTTGTTTT
Phenotype
MIM: 611176
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CCDC175 PubMed Links

Gene Details

Gene
CCDC175
Gene Name
coiled-coil domain containing 175
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164399.1 3444 Intron NP_001157871.1
XM_011537120.2 3444 UTR 3 XP_011535422.1
XM_011537122.2 3444 UTR 3 XP_011535424.1
XM_011537124.2 3444 UTR 3 XP_011535426.1
XM_011537125.2 3444 Intron XP_011535427.1
XM_011537130.2 3444 Intron XP_011535432.1
XM_017021621.1 3444 Intron XP_016877110.1
Gene
JKAMP
Gene Name
JNK1/MAPK8-associated membrane protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098625.2 3444 UTR 3 NP_001092095.1
NM_001284201.1 3444 UTR 3 NP_001271130.1
NM_001284202.1 3444 UTR 3 NP_001271131.1
NM_001284203.1 3444 UTR 3 NP_001271132.1
NM_001284204.1 3444 UTR 3 NP_001271133.1
NM_016475.4 3444 UTR 3 NP_057559.2
XM_006720171.3 3444 UTR 3 XP_006720234.1
XM_017021366.1 3444 UTR 3 XP_016876855.1

View Full Product Details