Product Details

SNP ID
rs4985830
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:21129976 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGGGTTGGTCATCTGTGCTACCTA[G/C]TTACTTTTTTCCAAAGGAAAACATG
Phenotype
MIM: 616160
Polymorphism
G/C, Transversion substitution
Allele Nomenclature
Literature Links
DHRS7B PubMed Links

Gene Details

Gene
DHRS7B
Gene Name
dehydrogenase/reductase 7B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015510.4 Intron NP_056325.2
XM_005256595.4 Intron XP_005256652.2
XM_005256596.3 Intron XP_005256653.1
XM_011523786.2 Intron XP_011522088.1
XM_017024425.1 Intron XP_016879914.1
XM_017024426.1 Intron XP_016879915.1

View Full Product Details