Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_000364.3 | Intron | NP_000355.2 | |||
NM_001001430.2 | Intron | NP_001001430.1 | |||
NM_001001431.2 | Intron | NP_001001431.1 | |||
NM_001001432.2 | Intron | NP_001001432.1 | |||
NM_001276345.1 | Intron | NP_001263274.1 | |||
NM_001276346.1 | Intron | NP_001263275.1 | |||
NM_001276347.1 | Intron | NP_001263276.1 | |||
XM_006711508.3 | Intron | XP_006711571.1 | |||
XM_006711509.3 | Intron | XP_006711572.1 | |||
XM_011509938.2 | Intron | XP_011508240.1 | |||
XM_011509939.1 | Intron | XP_011508241.1 | |||
XM_011509940.2 | Intron | XP_011508242.1 | |||
XM_011509941.2 | Intron | XP_011508243.1 | |||
XM_011509942.2 | Intron | XP_011508244.1 | |||
XM_011509943.2 | Intron | XP_011508245.1 | |||
XM_011509944.2 | Intron | XP_011508246.1 | |||
XM_011509946.1 | Intron | XP_011508248.1 | |||
XM_017002216.1 | Intron | XP_016857705.1 | |||
XM_017002217.1 | Intron | XP_016857706.1 |