Product Details
- SNP ID
-
rs2042468
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:24175577 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- CGAGTCCGTTTCTTTAATGGAGGTG[A/T]GATAACGTGGCGTTTTGAGACGTCA
- Phenotype
-
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
FAM228A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116450336,rs75121759] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FAM228A
- Gene Name
- family with sequence similarity 228 member A
- Gene
- FAM228B
- Gene Name
- family with sequence similarity 228 member B
There are no transcripts associated with this gene.
View Full Product Details