Product Details
- SNP ID
-
rs1835501
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:42746900 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GAAAAATTGGCATAGAAGGGACCTG[C/T]CTTACTGTAATAAAAGCTATCTATT
- Phenotype
-
MIM: 609592
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
RIT2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74364276] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RIT2
- Gene Name
- Ras like without CAAX 2
View Full Product Details