Product Details

SNP ID
rs71570637
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:63277040 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACACCAACTTACGTTATCAACTGA[C/T]TTGTAATCTATGGCTGAATATTTTC
Phenotype
MIM: 611470
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LGSN PubMed Links

Gene Details

Gene
LGSN
Gene Name
lengsin, lens protein with glutamine synthetase domain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143940.1 4124 UTR 3 NP_001137412.1
NM_016571.2 4124 UTR 3 NP_057655.2
XM_011535889.2 4124 Intron XP_011534191.1
XM_011535892.2 4124 Intron XP_011534194.1
XM_017010929.1 4124 Intron XP_016866418.1
XM_017010930.1 4124 Intron XP_016866419.1
XM_017010931.1 4124 Intron XP_016866420.1

View Full Product Details