Product Details

SNP ID
rs380988
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45867455 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTATACATTAACAATCTGTTTACAG[C/T]CAGTAGAGCTGTTCAAACTGTTTTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZSWIM3 PubMed Links

Gene Details

Gene
ZSWIM3
Gene Name
zinc finger SWIM-type containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080752.3 Intron NP_542790.2

View Full Product Details