Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001003676.2 | Intron | NP_001003676.1 | |||
NM_001003677.2 | Intron | NP_001003677.1 | |||
NM_001003678.2 | Intron | NP_001003678.1 | |||
NM_001278222.1 | Intron | NP_001265151.1 | |||
NM_024113.4 | Intron | NP_077018.1 | |||
XM_006718315.1 | Intron | XP_006718378.1 | |||
XM_011520364.2 | Intron | XP_011518666.1 | |||
XM_011520365.2 | Intron | XP_011518667.1 | |||
XM_017018276.1 | Intron | XP_016873765.1 | |||
XM_017018277.1 | Intron | XP_016873766.1 | |||
XM_017018278.1 | Intron | XP_016873767.1 | |||
XM_017018279.1 | Intron | XP_016873768.1 | |||
XM_017018280.1 | Intron | XP_016873769.1 | |||
XM_017018281.1 | Intron | XP_016873770.1 | |||
XM_017018282.1 | Intron | XP_016873771.1 |